4-tert-Butylbenzaldehyde 95%

Supplier: Ambeed
Danger

Synonyms: 4-(tert-Butyl)benzaldehyde

A426354-1000G A426354-100G A426354-10G A426354-25G A426354-500G
76804-206EA 119.89 USD
76804-206 76804-208 76804-210 76804-212 76804-214
4-tert-Butylbenzaldehyde 95%
4-tert-Butylbenzaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₁₄O
MW: 162.23 g/mol
MDL Number: MFCD00035742
CAS Number: 939-97-9
UN: 3082
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR