(2,4,6-Trimethylphenyl)acetonitrile 98%

Supplier: Ambeed
Warning

A426044-1G A426044-25G A426044-5G
76804-200EA 20 USD
76804-200 76804-202 76804-204
(2,4,6-Trimethylphenyl)acetonitrile 98%
(2,4,6-Trimethylphenyl)acetonitrile

Precaución: for Research Use Only

Formula: C₁₁H₁₃N
MDL Number: MFCD00013815
CAS Number: 34688-71-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR