Geranylacetone 98% (mixture of isomers)

Supplier: Ambeed
Warning

Synonyms: (E)-6,10-Dimethylundeca-5,9-dien-2-one

A423915-25G A423915-5G
76804-174EA 23.34 USD
76804-174 76804-176
Geranylacetone 98% (mixture of isomers)
Geranylacetone

Precaución: for Research Use Only

Formula: C₁₃H₂₂O
MW: 194.31 g/mol
MDL Number: MFCD00008910
CAS Number: 3796-70-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR