Ethyl-5-bromo-1-benzofuran-2-carboxylate 97%

Supplier: Ambeed
Warning

A423483-1G A423483-250MG A423483-25G A423483-5G
76804-158EA 11 USD
76804-158 76804-160 76804-162 76804-164
Ethyl-5-bromo-1-benzofuran-2-carboxylate 97%
Ethyl-5-bromo-1-benzofuran-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₉BrO₃
MDL Number: MFCD02667598
CAS Number: 84102-69-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR