3-(4-Thiazolyl)-L-alanine 95%

Supplier: Ambeed
Warning

Synonyms: H-3-(4-Thiazolyl)-Ala-OH, (S)-2-Amino-3-(thiazol-4-yl)propanoic acid

A422531-1G A422531-250MG
76804-144EA 145.35 USD
76804-144 76804-146
3-(4-Thiazolyl)-L-alanine 95%
3-(4-Thiazolyl)-L-alanine

Precaución: for Research Use Only

Formula: C₆H₈N₂O₂S
MW: 172.202 g/mol
MDL Number: MFCD00079677
CAS Number: 119433-80-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR