Lithium chloride monohydrate 99%
Supplier: Ambeed
|
Warning
|
Synonyms:
Lithium chloride hydrate, Hydrochloric acid lithium salt monohydrate
A420045-1000G
A420045-100G
A420045-25G
A420045-500G
76804-134EA
130.5
USD
76804-134
76804-136
76804-138
76804-140
Lithium chloride monohydrate 99%
Lithium chloride monohydrate
Precaución: for Research Use Only
Formula:
ClLi·1H₂O MW: 60.41 g/mol |
MDL Number:
MFCD00149764 CAS Number: 16712-20-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...