Diphenyl sulfide 98%

Supplier: Ambeed
Warning

A418779-100G A418779-10G A418779-25G A418779-500G
76804-126EA 35 USD
76804-126 76804-128 76804-130 76804-132
Diphenyl sulfide 98%
Diphenyl sulphide

Precaución: for Research Use Only

Formula: C₁₂H₁₀S
MDL Number: MFCD00003064
CAS Number: 139-66-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR