Di(N-succinimidyl) sebacate 98%

Supplier: Ambeed
Warning

Synonyms: Sebacic acid bis(N-succinimidyl) ester, Di(N-succinimidyl) Decanedioate, Sebacic acid Di(N-succinimidyl) ester, Decanedioic acid Di(N-succinimidyl) ester, Bis(2,5-dioxopyrrolidin-1-yl) decanedioate

A418472-5G A418472-25G A418472-1G
76804-124EA 73.21 USD
76804-124 77601-812 77626-838
Di(N-succinimidyl) sebacate 98%
Di(N-succinimidyl) sebacate

Precaución: for Research Use Only

Formula: C₁₈H₂₄N₂O₈
MW: 396.40 g/mol
MDL Number: MFCD00216005
CAS Number: 23024-29-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR