Imidazo[1,2-b]pyridazine-2-carboxylic acid 90%

Supplier: Ambeed
Warning

A418178-100MG A418178-1G A418178-250MG
76804-118EA 50.93 USD
76804-118 76804-120 76804-122
Imidazo[1,2-b]pyridazine-2-carboxylic acid 90%
Imidazo[1,2-b]pyridazine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₇H₅N₃O₂
MW: 163.1300 g/mol
MDL Number: MFCD08235229
CAS Number: 160911-42-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR