Ethyl 3-oxo-3-[4-(trifluoromethyl)phenyl]propanoate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl [4-(trifluoromethyl)benzoyl]acetate, Ethyl 3-oxo-3-(4-(trifluoromethyl)phenyl)propanoate

A417282-10G A417282-25G A417282-1G
76804-114EA 192.03 USD
76804-114 76804-116 77572-300
Ethyl 3-oxo-3-[4-(trifluoromethyl)phenyl]propanoate 98%
Ethyl 3-oxo-3-[4-(trifluoromethyl)phenyl]propanoate

Precaución: for Research Use Only

Formula: C₁₂H₁₁F₃O₃
MW: 260.209 g/mol
MDL Number: MFCD03424818
CAS Number: 106263-53-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR