(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(5-bromothiophen-2-yl)propanoic acid 96%

Supplier: Ambeed
Warning

A416191-100MG A416191-250MG
76804-110EA 40.32 USD
76804-110 76804-112
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(5-bromothiophen-2-yl)propanoic acid 96%
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(5-bromothiophen-2-yl)propanoic acid

Precaución: for Research Use Only

Formula: C₂₂H₁₈BrNO₄S
MW: 472.35 g/mol
MDL Number: MFCD01311745
CAS Number: 220497-50-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR