Tris(2,3-dibromopropyl)isocyanurate 98%

Supplier: Ambeed
Warning

Synonyms: 1,3,5-Tris(2,3-dibromopropyl)-1,3,5-triazinane-2,4,6-trione, Isocyanuric acid Tris(2,3-dibromopropyl) Ester

A415476-100G A415476-25G A415476-500G
76804-104EA 8 USD
76804-104 76804-106 76804-108
Tris(2,3-dibromopropyl)isocyanurate 98%
Tris(2,3-dibromopropyl)isocyanurate

Precaución: for Research Use Only

Formula: C₁₂H₁₅Br₆N₃O₃
MW: 728.69 g/mol
MDL Number: MFCD00010290
CAS Number: 52434-90-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR