(S)-2-Amino-5-phenylpentanoic acid 97%

Supplier: Ambeed
Warning

A415288-1G A415288-250MG A415288-5G
76804-098EA 41.38 USD
76804-098 76804-100 76804-102
(S)-2-Amino-5-phenylpentanoic acid 97%
(S)-2-Amino-5-phenylpentanoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₅NO₂
MW: 193.24 g/mol
MDL Number: MFCD04117835
CAS Number: 62777-25-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR