1-Naphthohydroxamic acid 98%

Supplier: Ambeed
Warning

A415125-100MG A415125-1G A415125-250MG A415125-5G
76804-092EA 35 USD
76804-092 76804-094 76804-096 77879-910
1-Naphthohydroxamic acid 98%
1-Naphthohydroxamic acid

Precaución: for Research Use Only

Formula: C₁₁H₉NO₂
MDL Number: MFCD00059546
CAS Number: 6953-61-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR