2,2,4,4-Tetramethyl-3-pentanone 98%

Supplier: Ambeed
Danger

Synonyms: Di-tert-butyl ketone, Hexamethylacetone

A414867-5G A414867-25G
76804-090EA 20.16 USD
76804-090 77572-292
2,2,4,4-Tetramethyl-3-pentanone 98%
2,2,4,4-Tetramethyl-3-pentanone

Precaución: for Research Use Only

Formula: C₉H₁₈O
MW: 142.24 g/mol
MDL Number: MFCD00008845
CAS Number: 815-24-7
UN: 1224
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR