Methyl 16-bromohexadecanoate 98%

Supplier: Ambeed
Danger

A414586-100MG A414586-1G A414586-250MG A414586-5G A414586-25G
76804-082EA 9 USD
76804-082 76804-084 76804-086 76804-088 77668-874
Methyl 16-bromohexadecanoate 98%
Methyl 16-bromohexadecanoate

Precaución: for Research Use Only

Formula: C₁₇H₃₃BrO₂
MDL Number: MFCD00040769
CAS Number: 26825-89-8
UN: 1760
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR