Ethyl-2-cyanoacrylate 95% stabilized

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-cyanoacrylate

Stabilization: Stabilised with 4-tert-butylpyrocatechol (TBC)

A411344-100G A411344-25G A411344-500G
76804-066EA 59.42 USD
76804-066 76804-068 76804-070
Ethyl-2-cyanoacrylate 95% stabilized
Ethyl-2-cyanoacrylate

Precaución: for Research Use Only

Formula: C₆H₇NO₂
MW: 125.1253 g/mol
MDL Number: MFCD00045615
CAS Number: 7085-85-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR