Methoxyacetyl chloride 97%
Supplier: Ambeed
|
Danger
|
A411219-10G
A411219-25G
A411219-5G
76804-060EA
11.67
USD
76804-060
76804-062
76804-064
Methoxyacetyl chloride 97%
Methoxyacetyl chloride
Precaución: for Research Use Only
Formula:
C₃H₅ClO₂ MW: 108.52 g/mol |
MDL Number:
MFCD00000728 CAS Number: 38870-89-2 UN: 2920 ADR: 8,3,Ⅱ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...