Isoquinoline-5-carbaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: Isoquinoline-5-carboxaldehyde

A410913-1G A410913-250MG A410913-5G
76804-054EA 30.77 USD
76804-054 76804-056 76804-058
Isoquinoline-5-carbaldehyde 98%
Isoquinoline-5-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₇NO
MW: 157.17 g/mol
MDL Number: MFCD03412483
CAS Number: 80278-67-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR