Benzyl-L-(+)-mandelate 98+%

Supplier: Ambeed
Warning

A410433-100G A410433-1G A410433-25G A410433-5G
76804-044EA 190 USD
76804-044 76804-046 76804-048 76804-050
Benzyl-L-(+)-mandelate 98+%
Benzyl-L-(+)-mandelate

Precaución: for Research Use Only

Formula: C₁₅H₁₄O₃
MDL Number: MFCD00274325
CAS Number: 62173-99-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR