3,4-dimethylthieno[2,3-b]thiophene-2,5-dicarboxylic acid 95%

Supplier: Ambeed
Warning

Synonyms: 3,4-Dimethylthieno[2,3-b]thiophene-2,5-dicarboxylic acid

A409907-100MG A409907-1G A409907-250MG
76804-030EA 46.68 USD
76804-030 76804-032 76804-034
3,4-dimethylthieno[2,3-b]thiophene-2,5-dicarboxylic acid 95%
3,4-dimethylthieno[2,3-b]thiophene-2,5-dicarboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₈O₄S₂
MW: 256.298 g/mol
MDL Number: MFCD00067989
CAS Number: 175202-55-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR