3,5-Di(pyridin-2-yl)-4H-1,2,4-triazol-4-amine 97%

Supplier: Ambeed
Warning

A408865-100MG A408865-1G A408865-250MG A408865-5G
76804-022EA 13.8 USD
76804-022 76804-024 76804-026 76804-028
3,5-Di(pyridin-2-yl)-4H-1,2,4-triazol-4-amine 97%
3,5-Di(pyridin-2-yl)-4H-1,2,4-triazol-4-amine

Precaución: for Research Use Only

Formula: C₁₂H₁₀N₆
MW: 238.24 g/mol
MDL Number: MFCD00075233
CAS Number: 1671-88-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR