Eosin Y (yellowish) ≥80% (dye content)
Supplier: Ambeed
|
Warning
|
A408452-100G
A408452-25G
A408452-500G
76804-012EA
21
USD
76804-012
76804-014
76804-016
Eosin Y (yellowish) ≥80% (dye content)
Eosin Y (yellowish)
Precaución: for Research Use Only
Formula:
C₂₀H₆Br₄Na₂O₅ |
MDL Number:
MFCD00005040 CAS Number: 17372-87-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...