Methoxy(dimethyl)-n-octylsilane 96% (by GC)

Supplier: Ambeed
Warning

Synonyms: Methoxydimethyl(octyl)silane

A407879-100G A407879-1G A407879-25G A407879-5G
76804-004EA 580.32 USD
76804-004 76804-006 76804-008 76804-010
Methoxy(dimethyl)-n-octylsilane 96% (by GC)
Methoxy(dimethyl)-n-octylsilane

Precaución: for Research Use Only

Formula: C₁₁H₂₆OSi
MW: 202.41 g/mol
MDL Number: MFCD00043322
CAS Number: 93804-29-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR