(±)-γ-Hexalactone 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(±)-γ-Caprolactone, γ-Caprolactone, 5-Ethyldihydrofuran-2(3H)-one, (±)-Hexan-4-olide
A405612-1000G
A405612-100G
A405612-10G
A405612-500G
A405612-5G
A405612-25G
76803-962EA
291.75
USD
76803-962
76803-964
76803-966
76803-968
76803-970
76841-654
(±)-γ-Hexalactone 98%
(±)-γ-Hexalactone
Precaución: for Research Use Only
Formula:
C₆H₁₀O₂ MW: 114.14 g/mol |
MDL Number:
MFCD00005401 CAS Number: 695-06-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...