Geranyl acetate 70%

Supplier: Ambeed
Warning

A404343-100G A404343-25G A404343-500G
76803-942EA 21 USD
76803-942 76803-944 76803-946
Geranyl acetate 70%
Geranyl acetate

Precaución: for Research Use Only

Formula: C₁₂H₂₀O₂
MDL Number: MFCD00015037
CAS Number: 105-87-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR