Ethyl-3-bromoimidazo[1,2-a]pyridine-5-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 3-bromoimidazo[1,2-a]pyridine-5-carboxylate

A401837-1G A401837-250MG
76803-932EA 147.47 USD
76803-932 76803-934
Ethyl-3-bromoimidazo[1,2-a]pyridine-5-carboxylate 97%
Ethyl 3-bromoimidazo[1,2-a]pyridine-5-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₉BrN₂O₂
MW: 269.0947 g/mol
MDL Number: MFCD10566747
CAS Number: 429690-42-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR