Ethyl chlorodifluoroacetate 97%

Supplier: Ambeed
Danger

Synonyms: Ethyl 2-chloro-2,2-difluoroacetate, Chlorodifluoroacetic ethyl ester

A399352-5G A399352-25G
76803-922EA 8 USD
76803-922 77879-867
Ethyl chlorodifluoroacetate 97%
Ethyl chlorodifluoroacetate

Precaución: for Research Use Only

Formula: C₄H₅ClF₂O₂
MW: 158.53 g/mol
MDL Number: MFCD00013662
CAS Number: 383-62-0
UN: 2924
ADR: 3,8 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR