Ethyl 3-amino-4-chloro-1-benzothiophene-2-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 3-amino-4-chlorobenzo[b]thiophene-2-carboxylate

A398481-1G A398481-250MG
76803-916EA 70.02 USD
76803-916 76803-918
Ethyl 3-amino-4-chloro-1-benzothiophene-2-carboxylate 97%
Ethyl 3-amino-4-chloro-1-benzothiophene-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₀ClNO₂S
MW: 255.721 g/mol
MDL Number: MFCD00661179
CAS Number: 67189-92-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR