Sodium (5R,6S)-6-((R)-1-hydroxyethyl)-7-oxo-3-((R)-tetrahydrofuran-2-yl)-4-thia-1-azabicyclo[3.2.0]hept-2-ene-2-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Faropenem sodium

A396403-1MG A396403-5MG
76803-902EA 32.89 USD
76803-902 76803-904
Sodium (5R,6S)-6-((R)-1-hydroxyethyl)-7-oxo-3-((R)-tetrahydrofuran-2-yl)-4-thia-1-azabicyclo[3.2.0]hept-2-ene-2-carboxylate 98%
Sodium (5R,6S)-6-((R)-1-hydroxyethyl)-7-oxo-3-((R)-tetrahydrofuran-2-yl)-4-thia-1-azabicyclo[3.2.0]hept-2-ene-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₄NNaO₅S
MW: 307.3 g/mol
MDL Number: MFCD01682061
CAS Number: 122547-49-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR