2-Formylcinnamic acid 97%

Supplier: Ambeed
Warning

A395822-1G A395822-250MG
76803-898EA 76.39 USD
76803-898 76803-900
2-Formylcinnamic acid 97%
2-Formylcinnamic acid

Precaución: for Research Use Only

Formula: C₁₀H₈O₃
MW: 176.169 g/mol
MDL Number: MFCD03840500
CAS Number: 130036-17-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR