L-3,4-Dihydroxyphenylalanine methyl ester hydrochloride 95%
Supplier: Ambeed
|
Warning
|
A394442-10G
A394442-100G
A394442-25G
76803-894EA
7.43
USD
76803-894
77572-202
77879-851
77879-852
L-3,4-Dihydroxyphenylalanine methyl ester hydrochloride 95%
L-3,4-Dihydroxyphenylalanine methyl ester hydrochloride
Precaución: for Research Use Only
Formula:
C₁₀H₁₄ClNO₄ |
MDL Number:
MFCD00038958 CAS Number: 1421-65-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...