4-(4-Methylphenyl)-4-oxobutyric acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Oxo-4-(p-tolyl)butanoic acid, 3-(4-Methylbenzoyl)propionic Acid, 4-(p-Tolyl)-4-oxobutyric Acid
A394352-1G
A394352-10G
A394352-25G
76803-892EA
5
USD
76803-892
77879-849
77879-850
4-(4-Methylphenyl)-4-oxobutyric acid 98%
4-(4-Methylphenyl)-4-oxobutyric acid
Precaución: for Research Use Only
Formula:
C₁₁H₁₂O₃ MW: 192.21 g/mol |
MDL Number:
MFCD00020541 CAS Number: 4619-20-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...