Iron(III) phosphate xhydrate 99+%

Supplier: Ambeed
Warning

A393511-100G A393511-25G A393511-1000G
76803-886EA 9 USD
76803-886 76803-888 77668-820
Iron(III) phosphate xhydrate 99+%
Iron(III) phosphate xhydrate

Precaución: for Research Use Only

Formula: FeH₂O₅P
MDL Number: MFCD00150236
CAS Number: 51833-68-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR