(2R,3S,4R,5R,6R)-5-Amino-2-(aminomethyl)-6-(((1R,2R,3S,4R,6S)-4,6-diamino-2-(((2S,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)oxy)-3-hydroxycyclohexyl)oxy)tetrahydro-2H-pyran-3,4-diol xsulfate 98%

Supplier: Ambeed
Danger

Synonyms: Ribostamycin sulphate

A391931-1G A391931-1MG A391931-25G A391931-5G
76803-870EA 26.53 USD
76803-870 76803-872 77626-790 77626-792
(2R,3S,4R,5R,6R)-5-Amino-2-(aminomethyl)-6-(((1R,2R,3S,4R,6S)-4,6-diamino-2-(((2S,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)oxy)-3-hydroxycyclohexyl)oxy)tetrahydro-2H-pyran-3,4-diol xsulfate 98%
(2R,3S,4R,5R,6R)-5-Amino-2-(aminomethyl)-6-(((1R,2R,3S,4R,6S)-4,6-diamino-2-(((2S,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)oxy)-3-hydroxycyclohexyl)oxy)tetrahydro-2H-pyran-3,4-diol xsulfate

Precaución: for Research Use Only

Formula: C₁₇H₃₆N₄O₁₄S
MW: 552.55 g/mol
MDL Number: MFCD23380549
CAS Number: 53797-35-6
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR