4-Nitro-1-naphthol 98%

Supplier: Ambeed
Warning

Synonyms: 4-Nitronaphthalen-1-ol, KAT8-IN-1, 1-Hydroxy-4-nitronaphthalene

A390905-1G A390905-5G A390905-100G A390905-25G
76803-854EA 6 USD
76803-854 76803-856 77626-786 77626-788
4-Nitro-1-naphthol 98%
4-Nitro-1-naphthol

Precaución: for Research Use Only

Formula: C₁₀H₇NO₃
MDL Number: MFCD02179392
CAS Number: 605-62-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR