5-Oxo-5,6,7,8-tetrahydronaphthalene-2-carboxylic acid 97%
Supplier: Ambeed
|
Warning
|
A390163-1G
A390163-250MG
A390163-5G
76803-836EA
67.9
USD
76803-836
76803-838
77668-800
5-Oxo-5,6,7,8-tetrahydronaphthalene-2-carboxylic acid 97%
5-Oxo-5,6,7,8-tetrahydronaphthalene-2-carboxylic acid
Precaución: for Research Use Only
Formula:
C₁₁H₁₀O₃ |
MDL Number:
MFCD01763813 CAS Number: 3470-46-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...