5-Amino-4-cyano-1-phenyl-1H-pyrazole 95%

Supplier: Ambeed
Warning

Synonyms: 5-Amino-1-phenyl-1H-pyrazole-4-carbonitrile

A389243-10G A389243-250MG
76803-832EA 89.12 USD
76803-832 76803-834
5-Amino-4-cyano-1-phenyl-1H-pyrazole 95%
5-Amino-4-cyano-1-phenyl-1H-pyrazole

Precaución: for Research Use Only

Formula: C₁₀H₈N₄
MW: 184.20 g/mol
MDL Number: MFCD00020730
CAS Number: 5334-43-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR