5-(4-Chlorophenyl)-2-furoic acid 96% (by HPLC)
Supplier: Ambeed
|
Warning
|
A388454-1G
A388454-250MG
A388454-5G
76803-822EA
48.81
USD
76803-822
76803-824
76803-826
5-(4-Chlorophenyl)-2-furoic acid 96% (by HPLC)
5-(4-Chlorophenyl)-2-furoic acid
Precaución: for Research Use Only
Formula:
C₁₁H₇ClO₃ |
MDL Number:
MFCD00216587 CAS Number: 41019-45-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...