Ethyl 2-amino-6-benzyl-4,5,6,7-tetrahydrothieno[2,3-c]pyridine-3-carboxylate hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: Tinoridine hydrochloride

A388367-10MG A388367-1MG A388367-25MG A388367-50MG A388367-5MG
76803-812EA 22.28 USD
76803-812 76803-814 76803-816 76803-818 76803-820
Ethyl 2-amino-6-benzyl-4,5,6,7-tetrahydrothieno[2,3-c]pyridine-3-carboxylate hydrochloride 97%
Ethyl 2-amino-6-benzyl-4,5,6,7-tetrahydrothieno[2,3-c]pyridine-3-carboxylate hydrochloride

Precaución: for Research Use Only

Formula: C₁₇H₂₁ClN₂O₂S
MW: 352.87 g/mol
MDL Number: MFCD01680536
CAS Number: 25913-34-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR