Fmoc-(N-γ-1-(4,4-dimethyl-2,6-dioxocyclohex-1-ylidene)ethyl)-L-α,γ-diaminobutyric acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Fmoc-Dab(Dde)-OH
A387609-1G
A387609-250MG
A387609-5G
76803-798EA
24
USD
76803-798
76803-800
76803-802
Fmoc-(N-γ-1-(4,4-dimethyl-2,6-dioxocyclohex-1-ylidene)ethyl)-L-α,γ-diaminobutyric acid 98%
Fmoc-(N-gamma-1-(4,4-dimethyl-2,6-dioxocyclohex-1-ylidene)ethyl)-L-alpha,gamma-diaminobutyric acid
Precaución: for Research Use Only
Formula:
C₂₉H₃₂N₂O₆ |
MDL Number:
MFCD11111552 CAS Number: 235788-61-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...