Fluorescamine 98%

Supplier: Ambeed
Warning

A386017-100MG A386017-10MG A386017-1G A386017-1MG A386017-250MG A386017-50MG
76803-758EA 41 USD
76803-758 76803-760 76803-762 76803-764 76803-766 76803-768
Fluorescamine 98%
Fluorescamine

Precaución: for Research Use Only

Formula: C₁₇H₁₀O₄
MDL Number: MFCD00005928
CAS Number: 38183-12-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR