Monomethyl itaconate 97%

Supplier: Ambeed
Warning

Synonyms: Itaconic Acid Monomethyl Ester, beta-Methyl Itaconate, 4-Methyl Methylenesuccinate, 4-Methoxy-2-methylene-4-oxobutanoic acid

A385351-100G A385351-1G A385351-25G A385351-5G
76803-750EA 145.35 USD
76803-750 76803-752 76803-754 76803-756
Monomethyl itaconate 97%
Monomethyl itaconate

Precaución: for Research Use Only

Formula: C₆H₈O₄
MW: 144.13 g/mol
MDL Number: MFCD00021713
CAS Number: 7338-27-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR