Hexadecanamide 98%

Supplier: Ambeed
Warning

A383540-100G A383540-100MG A383540-10G A383540-1G A383540-250MG A383540-25G A383540-5G
76803-736EA 139 USD
76803-736 76803-738 76803-740 76803-742 76803-744 76803-746 76803-748
Hexadecanamide 98%
Hexadecanamide

Precaución: for Research Use Only

Formula: C₁₆H₃₃NO
MDL Number: MFCD00025534
CAS Number: 629-54-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR