Dichloro(1,5-cyclooctadiene)platinum(II) 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
1,5-Cyclooctadieneplatinum(II) Dichloride
A381753-100MG
A381753-10G
A381753-1G
A381753-250MG
A381753-5G
76803-726EA
12.74
USD
76803-726
76803-728
77572-136
77601-678
77601-680
Dichloro(1,5-cyclooctadiene)platinum(II) 98%
Dichloro(1,5-cyclooctadiene)platinum(II)
Precaución: for Research Use Only
Formula:
C₈H₁₂Cl₂Pt MW: 374.17 g/mol |
MDL Number:
MFCD00012413 CAS Number: 12080-32-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...