Dimethyl 4-methoxyisophthalate 95+%

Supplier: Ambeed
Warning

A381067-1G A381067-250MG A381067-5G
76803-720EA 23 USD
76803-720 76803-722 76803-724
Dimethyl 4-methoxyisophthalate 95+%
Dimethyl 4-methoxyisophthalate

Precaución: for Research Use Only

Formula: C₁₁H₁₂O₅
MDL Number: MFCD06204071
CAS Number: 22955-73-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR