EDTA tripotassium salt dihydrate 99+%

Supplier: Ambeed
Warning

Synonyms: Ethylenedinitrilotetra-acetic acid tripotassium salt dihydrate, Tripotassium hydrogen ethylenediaminetetraacetate dihydrate, Ethylenediaminetetra-acetic acid tripotassium salt dihydrate, Potassium 2,2'-((2-((carboxylatomethyl)(carboxymethyl)amino)ethyl)azanediyl)diacetate dihydrate, Edetic acid tripotassium salt dihydrate

A380471-1000G A380471-100G A380471-25G A380471-500G
76803-706EA 123 USD
76803-706 76803-708 76803-710 76803-712
EDTA tripotassium salt dihydrate 99+%
EDTA tripotassium salt dihydrate

Precaución: for Research Use Only

Formula: C₁₀H₁₃K₃N₂O₈·2H₂O
MW: 442.55 g/mol
MDL Number: MFCD00149672
CAS Number: 65501-24-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR