cis-3-Hexenyl isovalerate 98+%

Supplier: Ambeed
Danger

Synonyms: (Z)-Hex-3-en-1-yl 3-methylbutanoate, Isovaleric Acid cis-3-Hexenyl Ester

A379705-25G A379705-5G
76803-702EA 28.65 USD
76803-702 76803-704
cis-3-Hexenyl isovalerate 98+%
cis-3-Hexenyl isovalerate

Precaución: for Research Use Only

Formula: C₁₁H₂₀O₂
MW: 184.28 g/mol
MDL Number: MFCD00036533
CAS Number: 35154-45-1
UN: 3272
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR