Diethylenetriaminepentaacetic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
N-Carboxymethyliminobis(ethylenenitrilo)tetra(acetic acid), Kestranal® Aza Pa, DTPA, Titriplex® V, Idranal V, Diethylenetriaminepentaacetic acid, 2,2',2'',2'''-((((Carboxymethyl)azanediyl)bis(ethane-2,1-diyl))bis(azanetriyl))tetraacetic acid, Diethylenetriamine-N,N,N',N'',N''-pentaacetic acid
A377951-1000G
A377951-100G
A377951-25G
A377951-500G
76803-690EA
49
USD
76803-690
76803-692
76803-694
76803-696
Diethylenetriaminepentaacetic acid 98%
Diethylenetriaminepentaacetic acid
Precaución: for Research Use Only
Formula:
C₁₄H₂₃N₃O₁₀ MW: 393.35 g/mol |
MDL Number:
MFCD00004289 CAS Number: 67-43-6 Merck Index: 13,07202 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...