Isoamyl n-octanoate 95%

Supplier: Ambeed
Warning

Synonyms: Isopentyl n-Octanoate, n-Octanoic Acid Isoamyl Ester, Isopentyl octanoate

A377688-100G A377688-25G A377688-500G
76803-678EA 22.28 USD
76803-678 76803-680 76803-682
Isoamyl n-octanoate 95%
Isoamyl n-octanoate

Precaución: for Research Use Only

Formula: C₁₃H₂₆O₂
MW: 214.35 g/mol
MDL Number: MFCD00048917
CAS Number: 2035-99-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR